siRNA Details
Gene Symbol: Bcl11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191683.1 | NP_001178612.1 | 4 | 820 | ||
XM_006251579.2 | XP_006251641.2 | 4 | 606 | |||
XM_006251586.2 | XP_006251648.1 | 3 | 941 | |||
XM_006251587.2 | XP_006251649.1 | 4 | 741 | |||
XM_006251588.2 | XP_006251650.1 | 4 | 1042 | |||
XM_006251591.2 | XP_006251653.1 | 3 | 2465 | |||
XM_006251594.2 | XP_006251656.1 | 4 | 1029 | |||
XM_008770430.1 | XP_008768652.1 | 4 | 1042 | |||
XM_008770431.1 | XP_008768653.1 | 4 | 897 | |||
GenBank mRNA | BC086607.1 | - | 4 | 929 | ||
BC158689.1 | - | 4 | 733 | 8375842 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001159289, NM_016707, NM_001159290, NM_001242934, AK043677, and others.
Gene Ontology Categories:
