siRNA Details
Gene Symbol: Efemp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012039.1 | NP_001012039.1 | 5 | 337 | ||
XM_008770456.1 | XP_008768678.1 | 5 | 437 | |||
XM_008770457.1 | XP_008768679.1 | 4 | 357 | |||
GenBank mRNA | BC079455.1 | - | 5 | 337 | 7121509 | |
D89730.1 | - | 3 | 233 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
