siRNA Details
Gene Symbol: Pcgf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107245.2 | NP_001100715.2 | 8 | 706 | ||
GenBank mRNA | BC167000.1 | - | 8 | 706 | 9031673 |

More Information
Other Species Targeted

Gene Ontology Categories:
