siRNA Details
Gene Symbol: Zfp385d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013992.1 | NP_001014014.1 | 6 | 1035 | ||
XM_008770513.1 | XP_008768735.1 | 5 | 3867 | |||
GenBank mRNA | BC079278.1 | - | 6 | 1035 | 7134172 |
