siRNA Details
Gene Symbol: Fezf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107251.1 | NP_001100721.1 | 4 | 1283 | ||
XM_006251737.2 | XP_006251799.1 | 6 | 1546 | |||
GenBank mRNA | BC168214.1 | - | 4 | 1113 | 7313369 |

More Information
Other Species Targeted

Gene Ontology Categories:
