siRNA Details
Gene Symbol: Nkiras1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107252.2 | NP_001100722.1 | 4 | 611 | ||
XM_006251698.2 | XP_006251760.1 | 3 | 585 | |||
XM_008770514.1 | XP_008768736.1 | 4 | 739 | |||
GenBank mRNA | BC169080.1 | - | 4 | 611 | 5623298 | |
FQ211880.1 | - | 496 |

More Information
Other Species Targeted

Gene Ontology Categories:
