siRNA Details
Gene Symbol: Rnase6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206815.1 | NP_996538.1 | 1 | 105 | ||
XM_006251871.2 | XP_006251933.1 | 2 | 174 | |||
XM_006251872.2 | XP_006251934.1 | 2 | 473 | |||
XM_008770517.1 | XP_008768739.1 | 3 | 318 | |||
XM_008770518.1 | XP_008768740.1 | 3 | 292 | |||
XM_008770519.1 | XP_008768741.1 | 2 | 192 | |||
XM_008770520.1 | XP_008768742.1 | 2 | 171 |
Gene Symbol: LOC103694852 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773787.1 | XP_008772009.1 | 2 | 174 | ||
XM_008773788.1 | XP_008772010.1 | 2 | 473 | |||
XM_008773789.1 | XP_008772011.1 | 3 | 321 | |||
XM_008773790.1 | XP_008772012.1 | 3 | 295 | |||
XM_008773791.1 | XP_008772013.1 | 2 | 195 | |||
XM_008773792.1 | XP_008772014.1 | 2 | 171 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
