siRNA Details
Gene Symbol: Rnase1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025116.1 | NP_001020287.1 | 1 | 211 | ||
XM_006251875.1 | XP_006251937.1 | 2 | 319 |
Gene Symbol: LOC103694853 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773795.1 | XP_008772017.1 | 319 |
