siRNA Details
Gene Symbol: Rpgrip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006251921.2 | XP_006251983.1 | 11, 12 | 1352 | ||
XM_008770631.1 | XP_008768853.1 | 11, 12 | 1320 | |||
XM_008770633.1 | XP_008768855.1 | 10, 11 | 1222 | |||
XM_008770634.1 | XP_008768856.1 | 11, 12 | 1349 | |||
XM_008770635.1 | XP_008768857.1 | 11, 12 | 1345 | |||
XM_008770636.1 | XP_008768858.1 | 11, 12 | 1345 | |||
GenBank mRNA | BC097397.1 | - | 1, 2 | 262 |
