siRNA Details
Gene Symbol: Ap5m1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030036.1 | NP_001025207.1 | 6, 7 | 1518 | ||
GenBank mRNA | BC099829.1 | - | 6, 7 | 1518 | 7933740 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ap5m1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030036.1 | NP_001025207.1 | 6, 7 | 1518 | ||
GenBank mRNA | BC099829.1 | - | 6, 7 | 1518 | 7933740 |