siRNA Details
Gene Symbol: Acin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170468.1 | NP_001163939.1 | 14 | 3008 | ||
XM_006251958.2 | XP_006252020.1 | 14 | 2941 | |||
XM_006251959.2 | XP_006252021.1 | 14 | 2938 | |||
XM_006251960.2 | XP_006252022.1 | 13 | 2857 | |||
XM_006251962.2 | XP_006252024.1 | 13 | 2666 | |||
XM_006251963.1 | XP_006252025.1 | 7 | 981 | |||
XM_006251964.1 | XP_006252026.1 | 7 | 763 | |||
GenBank mRNA | BC091320.1 | - | 5 | 501 | ||
CA512047.1 | - | 6 | 593 | |||
EV770592.1 | - | 1 | 63 |

More Information
Other Species Targeted

Gene Ontology Categories:
