siRNA Details
Gene Symbol: Acin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170468.1 | NP_001163939.1 | 7 | 2039 | ||
XM_006251958.2 | XP_006252020.1 | 7 | 2008 | |||
XM_006251959.2 | XP_006252021.1 | 7 | 2008 | |||
XM_006251960.2 | XP_006252022.1 | 6 | 1888 | |||
XM_006251962.2 | XP_006252024.1 | 6 | 1697 | |||
GenBank mRNA | CB744719.1 | - | 2 | 308 | ||
FM029308.1 | - | 1 | 19 |
