siRNA Details
Gene Symbol: Cpne6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191113.1 | NP_001178042.1 | 6 | 634 | ||
GenBank mRNA | AM746509.1 | - | 5 | 521 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cpne6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191113.1 | NP_001178042.1 | 6 | 634 | ||
GenBank mRNA | AM746509.1 | - | 5 | 521 |