siRNA Details
Gene Symbol: Dcaf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009686.2 | NP_001009686.1 | 12 | 1389 | ||
XM_006251971.2 | XP_006252033.1 | 13 | 1724 | |||
GenBank mRNA | BC081957.1 | - | 13 | 1594 | ||
BC087110.1 | - | 11 | 1170 | 7193336 | ||
BC091364.1 | - | 12 | 1389 | 7319156 | ||
FQ222851.1 | - | 13 | 1690 |

More Information
Other Species Targeted

Gene Ontology Categories:
