siRNA Details
Gene Symbol: Pspc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025672.1 | NP_001020843.1 | 1 | 342 | ||
XM_006252065.2 | XP_006252127.1 | 1 | 384 | |||
GenBank mRNA | BC099204.1 | - | 1 | 342 | 7384173 |
