siRNA Details
Gene Symbol: Zmym2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770810.1 | XP_008769032.1 | 3 | 579 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zmym2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770810.1 | XP_008769032.1 | 3 | 579 |