siRNA Details
Gene Symbol: Lats2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107267.1 | NP_001100737.1 | 7 | 2847 | ||
XM_008770748.1 | XP_008768970.1 | 8 | 2892 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lats2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107267.1 | NP_001100737.1 | 7 | 2847 | ||
XM_008770748.1 | XP_008768970.1 | 8 | 2892 |