siRNA Details
Gene Symbol: Wdfy2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107269.1 | NP_001100739.1 | 2 | 429 | ||
XM_006252177.2 | XP_006252239.1 | 3 | 465 | |||
XM_008770751.1 | XP_008768973.1 | 4 | 2342 | |||
XM_008770752.1 | XP_008768974.1 | 5 | 1935 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_175546, AK086275, BC116632, AK169499, AK151817, and others.
Gene Ontology Categories:
