siRNA Details
Gene Symbol: Kif13b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_213626.1 | NP_998791.1 | 7, 8 | 583 | ||
XM_008770753.1 | XP_008768975.1 | 7, 8 | 805 | |||
XM_008770754.1 | XP_008768976.1 | 7, 8 | 805 | |||
XM_008770755.1 | XP_008768977.1 | 7, 8 | 805 | |||
GenBank mRNA | AJ605719.1 | - | 7, 8 | 583 |
