siRNA Details
Gene Symbol: Polr3d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031653.1 | NP_001026823.1 | 7, 8 | 961 | ||
XM_006252272.2 | XP_006252334.1 | 7, 8 | 961 | |||
GenBank mRNA | BC099167.1 | - | 7, 8 | 961 | 7382125 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
