siRNA Details
Gene Symbol: Enox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770914.1 | XP_008769136.1 | 16 | 3690 | ||
XM_008770915.1 | XP_008769137.1 | 16 | 3687 | |||
XM_008770916.1 | XP_008769138.1 | 15 | 3633 | |||
GenBank mRNA | FQ222793.1 | - | 3315 |

More Information
Other Species Targeted

Gene Ontology Categories:
