siRNA Details
Gene Symbol: Pcdh9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191688.1 | NP_001178617.1 | 1 | 1531 | ||
XM_008770867.1 | XP_008769089.1 | 2 | 2170 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001271798, NM_001271800, NM_001271799, NM_001081377, AK147451, and others.
Gene Ontology Categories:
