siRNA Details
Gene Symbol: Pcdh9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191688.1 | NP_001178617.1 | 1 | 2730 | ||
XM_008770867.1 | XP_008769089.1 | 2 | 3369 | |||
GenBank mRNA | DQ863132.1 | - | 1 | 217 |

More Information
Other Species Targeted

Gene Ontology Categories:
