siRNA Details
Gene Symbol: Pibf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006252404.2 | XP_006252466.1 | 7, 8 | 1176 | ||
XM_006252405.2 | XP_006252467.1 | 7, 8 | 1129 | |||
XM_006252406.2 | XP_006252468.1 | 7, 8 | 961 | |||
XM_006252407.2 | XP_006252469.1 | 7, 8 | 1176 | |||
XM_224451.8 | XP_224451.3 | 7, 8 | 1002 | |||
GenBank mRNA | FQ210517.1 | - | 1164 |
