siRNA Details
Gene Symbol: Ipo5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006252478.2 | XP_006252540.1 | 11 | 1126 | ||
XM_224534.8 | XP_224534.7 | 12 | 1372 |

More Information
Other Species Targeted

Gene Ontology Categories:
