siRNA Details
Gene Symbol: Farp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107287.1 | NP_001100757.1 | 11 | 1142 | ||
XM_006252472.2 | XP_006252534.1 | 12 | 1706 | |||
XM_006252473.2 | XP_006252535.1 | 12 | 1752 | |||
XM_006252475.2 | XP_006252537.1 | 12 | 1706 |
