siRNA Details
Gene Symbol: Clybl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100685.1 | NP_001094155.1 | 3 | 276 | ||
GenBank mRNA | BC088243.1 | - | 3 | 256 | ||
FM063772.1 | - | 3 | 276 | |||
FQ214276.1 | - | 2 | 61 |

More Information
Other Species Targeted

Gene Ontology Categories:
