siRNA Details
Gene Symbol: Clybl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100685.1 | NP_001094155.1 | 2 | 199 | ||
XM_006252508.2 | XP_006252570.1 | 2 | 217 | |||
GenBank mRNA | BC088243.1 | - | 2 | 179 | ||
FM063772.1 | - | 2 | 199 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
