siRNA Details
Gene Symbol: Pde12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013998.2 | NP_001014020.2 | 2, 3 | 1286 | ||
GenBank mRNA | AY387060.1 | - | 2, 3 | 1286 | ||
BC079396.1 | - | 2, 3 | 1470 | 7112419 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
