siRNA Details
Gene Symbol: Itih1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107291.2 | NP_001100761.2 | 4 | 356 | ||
XM_008770995.1 | XP_008769217.1 | 4 | 401 | |||
GenBank mRNA | BC166831.1 | - | 4 | 356 | 8374222 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
