siRNA Details
Gene Symbol: Capn7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030037.1 | NP_001025208.1 | 9, 10 | 1196 | ||
XM_006252608.2 | XP_006252670.2 | 10, 11 | 1429 | |||
XM_008770997.1 | XP_008769219.1 | 9, 10 | 2000 | |||
GenBank mRNA | BC099770.1 | - | 9, 10 | 1196 | 7932056 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
