siRNA Details
Gene Symbol: Oxnad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107295.1 | NP_001100765.1 | 7 | 927 | ||
XM_006252623.2 | XP_006252685.1 | 7 | 1028 | |||
XM_006252624.2 | XP_006252686.1 | 8 | 1135 | |||
XM_006252625.2 | XP_006252687.1 | 8 | 1121 | |||
XM_006252626.2 | XP_006252688.1 | 7 | 1014 | |||
XM_008771000.1 | XP_008769222.1 | 8 | 1216 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
