siRNA Details
Gene Symbol: Arhgap22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107297.1 | NP_001100767.1 | 4, 5 | 633 | ||
XM_006252744.2 | XP_006252806.1 | 9, 10 | 1441 | |||
XM_006252749.2 | XP_006252811.1 | 6, 7 | 770 | |||
XM_006252750.2 | XP_006252812.1 | 6, 7 | 1350 | |||
XM_006252751.2 | XP_006252813.1 | 5, 6 | 1211 | |||
XM_008771098.1 | XP_008769320.1 | 8, 9 | 1496 |
