siRNA Details
Gene Symbol: Arhgap22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107297.1 | NP_001100767.1 | 5 | 999 | ||
XM_006252744.2 | XP_006252806.1 | 10 | 1807 | |||
XM_006252749.2 | XP_006252811.1 | 7 | 1136 | |||
XM_006252750.2 | XP_006252812.1 | 7 | 1716 | |||
XM_006252751.2 | XP_006252813.1 | 6 | 1577 | |||
XM_008771098.1 | XP_008769320.1 | 9 | 1862 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
