siRNA Details
Gene Symbol: Mmrn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751550.3 | XP_003751598.2 | 6 | 1397 | ||
GenBank mRNA | FQ218167.1 | - | 6 | 1332 |

More Information
Other Species Targeted

Gene Ontology Categories:
