siRNA Details
Gene Symbol: Sdccag3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013135.2 | NP_001013153.2 | 8, 9 | 1169 | ||
XM_006233681.2 | XP_006233743.1 | 7, 8 | 1079 | |||
XM_006233682.2 | XP_006233744.1 | 7, 8 | 998 | |||
XM_006233683.2 | XP_006233745.1 | 8, 9 | 1058 | |||
XM_006233684.2 | XP_006233746.1 | 6, 7 | 911 | |||
XM_006233685.2 | XP_006233747.1 | 6, 7 | 935 | |||
GenBank mRNA | BC088296.1 | - | 2, 3 | 109 | 7322728 | |
BC168170.1 | - | 3, 4 | 2456 | 9025284 |
Gene Symbol: LOC100363321 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006252792.1 | XP_006252854.1 | 1 | 934 |
Gene Symbol: RGD1562289 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_576917.5 | XP_576917.1 | 1 | 1003 |
