siRNA Details
Gene Symbol: Arrdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107303.1 | NP_001100773.1 | 7, 8 | 1523 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Arrdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107303.1 | NP_001100773.1 | 7, 8 | 1523 |