siRNA Details
Gene Symbol: Sh2d4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012048.1 | NP_001012048.1 | 6, 7 | 1059 | ||
XM_006253032.2 | XP_006253094.1 | 6, 7 | 1072 | |||
GenBank mRNA | BC079100.1 | - | 6, 7 | 1059 | 7124466 |
