siRNA Details
Gene Symbol: Ddx60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771220.1 | XP_008769442.1 | 35 | 4825 |
Gene Symbol: LOC100911190 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771241.1 | XP_008769463.1 | 24 | 4606 |
