siRNA Details
Gene Symbol: Gpm6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178105.2 | NP_835206.1 | 2 | 241 | ||
XM_006253093.2 | XP_006253155.1 | 3 | 1077 | |||
GenBank mRNA | AB089242.1 | - | 2 | 220 | ||
BC088862.1 | - | 2 | 241 | 7313006 | ||
FQ211804.1 | - | 3 | 696 |

More Information
Other Species Targeted

Gene Ontology Categories:
