siRNA Details
Gene Symbol: Stox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134863.1 | NP_001128335.1 | 3 | 3409 | ||
XM_006253151.2 | XP_006253213.2 | 3 | 2692 | |||
XM_006253152.2 | XP_006253214.1 | 3 | 2120 | |||
XM_006253153.2 | XP_006253215.1 | 3 | 2500 | |||
XM_008771258.1 | XP_008769480.1 | 3 | 2260 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
