siRNA Details
Gene Symbol: Enpp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107311.1 | NP_001100781.1 | 5 | 756 | ||
XM_006253156.2 | XP_006253218.1 | 5 | 774 | |||
XM_008771259.1 | XP_008769481.1 | 5 | 774 | |||
GenBank mRNA | BC158772.1 | - | 5 | 736 | 7128690 |
Gene Symbol: LOC100911549 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767456.1 | XP_008765678.1 | 4 | 513 |
