siRNA Details
Gene Symbol: Unc5d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107319.2 | NP_001100789.2 | 3, 4 | 460 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_153135, AJ487854, BC144784, BC144783, FJ824810, and others.
Gene Ontology Categories:
