siRNA Details
Gene Symbol: Pomk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024883.1 | NP_001020054.1 | 5 | 781 | ||
XM_008771353.1 | XP_008769575.1 | 3 | 555 | |||
GenBank mRNA | BC097466.1 | - | 5 | 781 | 7457667 | |
FQ211927.1 | - | 5 | 770 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_029037, AK043620, AK032677, AK138952, BC027296, and others.
Gene Ontology Categories:
