siRNA Details
Gene Symbol: Slc25a15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047880.1 | NP_001041345.1 | 6 | 735 | ||
XM_006253350.2 | XP_006253412.1 | 6 | 1171 | |||
XM_006253351.2 | XP_006253413.1 | 6 | 1562 | |||
XM_006253352.2 | XP_006253414.1 | 6 | 1388 | |||
XM_006253353.2 | XP_006253415.1 | 5 | 2216 | |||
GenBank mRNA | AY325139.1 | - | 6 | 735 |

More Information
Other Species Targeted

Gene Ontology Categories:
