siRNA Details
Gene Symbol: Nek3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006253359.2 | XP_006253421.1 | 12, 13 | 1281 | ||
XM_006253360.2 | XP_006253422.1 | 12, 13 | 1272 | |||
XM_006253361.2 | XP_006253423.1 | 12, 13 | 1278 | |||
XM_006253362.2 | XP_006253424.1 | 12, 13 | 1275 | |||
XM_006253363.1 | XP_006253425.1 | 10, 11 | 1060 | |||
XM_224971.7 | XP_224971.5 | 12, 13 | 1272 |
