siRNA Details
Gene Symbol: Agpat5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134744.1 | NP_001128216.1 | 8 | 916 | ||
XM_006253381.2 | XP_006253443.1 | 8 | 1188 | |||
GenBank mRNA | BC168222.1 | - | 8 | 916 | 7323695 |
