siRNA Details
Gene Symbol: Agpat5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134744.1 | NP_001128216.1 | 5 | 522 | ||
XM_006253381.2 | XP_006253443.1 | 5 | 794 | |||
GenBank mRNA | BC168222.1 | - | 5 | 522 | 7323695 |

More Information
Other Species Targeted

Gene Ontology Categories:
