siRNA Details
Gene Symbol: Atp11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107324.2 | NP_001100794.2 | 12 | 1392 | ||
XM_006253445.2 | XP_006253507.1 | 12 | 1347 | |||
XM_006253446.2 | XP_006253508.1 | 12 | 1347 | |||
XM_006253447.2 | XP_006253509.1 | 12 | 1347 | |||
XM_006253448.2 | XP_006253510.1 | 12 | 1347 |
