siRNA Details
Gene Symbol: Proz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006253473.1 | XP_006253535.1 | 5, 6 | 617 | ||
XM_006253474.1 | XP_006253536.1 | 4, 5 | 503 | |||
GenBank mRNA | BC160868.1 | - | 5, 6 | 529 | ||
FQ209855.1 | - | 5, 6 | 531 |
